Industrial gloves, ABERDEEN, TRIUS®
Leverandør: TRIUS SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABERDEEN/10 ABERDEEN/11 ABERDEEN/8 ABERDEEN/9
TRIUABERDEEN/10EA 1150 DKK
TRIUABERDEEN/10 TRIUABERDEEN/11 TRIUABERDEEN/8 TRIUABERDEEN/9
Industrial gloves, ABERDEEN, TRIUS®
Handsker
Impact and oil-resistant, cut and sewn synthetic gloves offer protection against shock, cuts, and fat. The ABERDEEN series solves multiple problems while ensuring high visibility.
  • High cut resistance
  • Impact protection
  • Palm and thumb reinforcement
  • Oil-proof
  • High visibility
ABERDEEN gloves are designed for use in wet and oily environments, with risks of cut or impact. This protective glove features a palm made of a 3-layer complex (synthetic leather/cut protection/nitrile coating textile), a back made of cut-resistant and oil­repellent high visibility textile with protection against shock, and a highly breathable Hyprene® wrist. They are washable and durable.
Certificeringer: EN 388:2016 (2X44EP)
Emballering: Bundle of 5 pairs.