BioSample cards
Leverandør: Ahlstrom-Munksjö
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8.460.0001.B-N 8.460.0012.A-N 8.460.0004.A-N 8.460.0004.B-N 8.460.0005.B-N 8.460.0038.A-N 8.294.0001.B-N 2.460.210297
BINZ8.460.0001.B-NEA 1560 DKK
BINZ8.460.0001.B-N BINZ8.460.0012.A-N BINZ8.460.0004.A-N BINZ8.460.0004.B-N BINZ8.460.0005.B-N BINZ8.460.0038.A-N BINZ8.294.0001.B-N BINZ2.460.210297
BioSample cards
Kliniske diagnose systemer Clinical Sample Collection/Preparation Biological Sample Collection/Preparation
These BioSample cards are designed for microvolume sampling, transport and ambient storage of biological samples. These cards are made of white and pure absorbent fibers and contain no wet-strength additives or chemicals. This feature ensures reproducible collection and minimise possible interferences for reliable analysis of targeted analytes like small molecules, proteins, nucleic acids or vitamins.
  • Reliable and reproducible sample collection
  • Even and uniform sample distribution
  • Customisable design and printing
  • Rigorous quality control
  • Each card has five/eight spots
General applications include protein-based analysis, lipid-based analysis, genomics, human identification and forensics. Indicated cards can also be used for HIV viral load testing and infectious disease screening (including Hepatitis B, C and Syphilis).
Certificeringer: Manufacturing according to ISO 13485:2016 and ISO 9001:2015 standards.
In Vitro Diagnostic medical device in compliance with the Directive 98/79/EC.
Bestillingsoplysninger: Drying racks are available to facilitate drying of the cards at ambient temperature.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904