1-Fluor-2,4-dinitrobenzen ≥99.0% (by GC), LiChropur™ derivatisation reagent, for HPLC, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 1,3-Dinitro-4-fluorobenzene, 2,4-Dinitrofluorobenzene, 1-Fluoro-2,4-dinitrobenzene
73177-10X1G 73177-1G
73177-10X1GEA 2280 DKK
73177-10X1G 73177-1G
1-Fluor-2,4-dinitrobenzen ≥99.0% (by GC), LiChropur™ derivatisation reagent, for HPLC, Supelco®
1-Fluor-2,4-dinitrobenzen
Formel: C₆H₃FN₂O₄
Molekylvægt: 186.1 g/mol
Kogepunkt: 296
MDL Number: MFCD00007056
CAS-nummer: 70-34-8
Merck Index: 13,04199
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=056f58ec8bfc32933b776d70d1403d6615b5323e&vid=056f58ec8bfc32933b776d70d1403d6615b5323e&prid=web&clid=SAPDAM