VWR®, Bottles, PET, Square, with Screw Cap
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
391-0629EA 649 DKK
391-0629 391-0628 391-0627 391-0631 391-0626 391-0648
VWR®, Bottles, PET, Square, with Screw Cap
Flasker Plastflasker
Square PET media bottles that are designed for packaging, storing, or transporting powder and liquid samples.
  • Square design allows efficient use of storage space
  • PET resin is lightweight, break-resistant and transparent
  • Bottles feature moulded-in graduations
  • EB Sterilisation Sterility Assurance Level (SAL) of 10⁻⁶ (ISO 11137)
  • Non pyrogenic
VWR® square PET media bottles are manufactured in a Class 100 000 cleanroom environment. Manufactured under EN ISO 13485:2016.
Certificeringer: Bottles meet the requirements of BSE/TSE, the BPA statement and heavy metals ROHS.
Emballering: Bottles are available individually wrapped, or in tray packs (triple-bagged PET bottles for cleanroom applications).
Advarsel: These bottles are not to be used for production packaging.