Methansulfonsyre ≥99.5% (by titrimetric analysis), LiChropur™ for HPLC, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
59510-10X1ML 59510-1ML
59510-10X1MLEA 1330 DKK
59510-10X1ML 59510-1ML
Methansulfonsyre ≥99.5% (by titrimetric analysis), LiChropur™ for HPLC, Supelco®
Methansulfonsyre
Formel: CH₃SO₃H
MDL Number: MFCD00007518
CAS-nummer: 75-75-2
UN nummer: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73a1de9e324b6249302c1b76b4dc74f87062a0a8&vid=a756f8d69b364437d6a5c58bf88c5d8f482fe32c&prid=web&clid=SAPDAM