Measuring cylinders, conical
Leverandør: BOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8660 8661 8662 8663
BOCH8660EA 241 DKK
BOCH8660 BOCH8661 BOCH8662 BOCH8663
Measuring cylinders, conical
Cylindere Målecylindre
Stainless steel 18/10.
  • With base and handle
  • Conical, graduated