CF®DI Hydrazide
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
92166 92164 92165 92184 92185
92166.EA 2190 DKK
92166. 92164. 92165. 92184. 92185.
CF®DI Hydrazide
Proteinkoblings- og mærkningssystemer
CF®DI dye hydrazides react with molecules containing aldehyde or ketone groups to form a hydrazone bond. The dyes are size- and charge-matched and do not significantly alter the molecular weight or pI of the labeled proteins. They are used for detection of protein carbonylation by fluorescence 2D-DIGE.
  • React with carbonyl moieties of aldehyde or ketone groups.
  • Size- and charge-matched, do not significantly alter the molecular weight or pI of the labeled protein.
  • For detection of protein carbonylation by fluorescence 2D-DIGE.
  • Available in several colors for detection of multiple samples on the same gel.
Also see our regular CF® Dye Hydrazides, available in a wide selection of colors.