Avantor® Partisil ODS2, HPLC Columns
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
P10ODS2-250AEA 0 DKK
P10ODS2-250A P10ODS2-250P
Avantor® Partisil ODS2, HPLC Columns
Kromatografikolonner
Avantor® Partisil® is one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity for preparative work.
  • Irregular porous silica
  • Convenient and easy to use: Hand tightened fittings
  • Increased column lifetime
The high carbon load C18 bonded silica phase makes it the most non-polar.
Advarsel: *Use with freestanding holder HI-161 and column coupler HI-081, please see "HPLC Guard Column Holders and Accessories"