Avantor® Partisil C8, HPLC Columns
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
P10C8-250AEA 5710 DKK
P10C8-250A P5C8-150AS P5C8-10C5 P10C8-150AS P5C8-100A P5C8-100AF P5C8-10C5-SK HICHP5C8-75A P5C8-100P P10C8-250P
Avantor® Partisil C8, HPLC Columns
Kromatografikolonner
Avantor® Partisil® was one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity.
  • Easy to use
  • Irregular porous silica
Avantor® Partisil® C8 is recommended for ion-pair chromatography.
Advarsel: * Use with freestanding holder HI-161 and column coupler HI-081, please see “HPLC Guard Column Holders and Accessories”.