VWR®, TLC Aluminium Plates, Silica, Unmodified Layers
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
551-0005EA 896 DKK
551-0005 551-0018 551-0020 551-0021 551-0032
VWR®, TLC Aluminium Plates, Silica, Unmodified Layers
TLC-plader
These TLC aluminium plates are suitable for easy and quick sample applications. They are flexible and can be cut to match different plate size requirements.
  • High sample throughput in short time
  • Pilot product for HPLC or Flash chromatography
  • Suitable for screening tests
Indicator: Manganese activated zinc silica with green fluorescence for short wave UV (254 nm).
Binder: Stable in almost all organic solvents.
Particle size: 5 to 17 µm