Supelco® Organic Reference Standard, (+)-Menthofuran
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
63661-1ML-F 63661-5ML-F
63661-1ML-FEA 950 DKK
63661-1ML-F 63661-5ML-F
Supelco® Organic Reference Standard, (+)-Menthofuran
Standarder Økologiske standarder
Advarsel: P264 - P270 - P273 - P301 + P312 - P302 + P352 - P305 + P351 + P338
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bfb513e03c75fef70f28c8bd737facd2b587566f&vid=bfb513e03c75fef70f28c8bd737facd2b587566f&prid=web&clid=SAPDAM
Bestil nu
Specifikationer
M-Clarity quality level MQ100
Synonym(s) (6R)-4,5,6,7-Tetrahydro-3,6-dimethylbenzofuran; (R)-3,6-Dimethyl-4,5,6,7-tetrahydrobenzofuran
Environmentally Preferable
Densitet 0,97 g/ml at 20 °C (lit.)
CAS-nr 17957-94-7
MDL No. MFCD00142332
Formel C₁₀H₁₄O
Molecular weight 150,22
Kogepunkt 204...206 °C (lit.)
Fareklassebeskrivelse H302 - H315 - H319 - H411
Shipping conditions STD
Storage conditions 2...8 °C
Opbevaringstemperatur 2…8 °C