VWR®, Cleanroom Gloves, Nitrile, Sterile, ISO 5
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
112-5431EA 4320 DKK
112-5431 112-5427 112-5428 112-5429 112-5430 112-5426
VWR®, Cleanroom Gloves, Nitrile, Sterile, ISO 5
Handsker
These soft nitrile cleanroom gloves are manufactured from acrylonitrile butadiene.
  • Ambidextrous, textured fingers and beaded cuff
  • Without natural rubber latex
  • Static-dissipative
  • Single-use only
  • Sterilisation by gamma irradiation to SAL 10⁻⁶
Suitable for use in ISO Class 5 (FED-STD-209E Class 100) or higher cleanrooms.
Certificeringer: EN ISO 374-1:2016, EN ISO 374-5:2016