Cleanroom wipes, pre-saturated, SATWipes®
Leverandør: CONTEC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SAT-C3-AC SW420192 SW420195
CONTSAT-C3-ACEA 4810 DKK
CONTSAT-C3-AC CONTSW420192 CONTSW420195
Cleanroom wipes, pre-saturated, SATWipes®
Papirservietter Servietter til Renrum Imprægneret
Non woven wipes pre-saturated with acetone or isopropanol.
  • Solvent-specific color-coded labeling for easy solvent identification
  • Wipes are dispensed from center-pull perforated rolls with self-closing lids for added convenience
  • Reusable canister is economical and solvent compatible
Contec's SATWipes® Solvent Wiping System consists of non woven pre-saturated wipes dispensed from a center-pull roll in a reusable canister. Custom engineered to meet a variety of needs, an assortment of fabrics, canisters and solvents are available. All fabrics andMore Product Information
Bestil nu
Specifikationer
Environmentally Preferable
Grundvægt 71 g/m²
Ekstraherbare stoffer - Med deioniseret vand 0,027 g/m²