Tetrabutylammoniumhydrogen sulfat ≥99.0% for ion pair chromatography, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Tetrabutylammonium bisulphate, Tetrabutylammonium hydrogensulphate
86853-10G-F 86853-50G-F 86853-10X1G-F
86853-10G-FEA 1180 DKK
86853-10G-F 86853-50G-F 86853-10X1G-F
Tetrabutylammoniumhydrogen sulfat ≥99.0% for ion pair chromatography, Supelco®
Tetrabutylammoniumhydrogen sulfat
Formel: (C₄H₉)₄NHSO₄
Molekylvægt: 339.54 g/mol
MDL Number: MFCD00011637
CAS-nummer: 32503-27-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ac945220fa3730d357572c05c3735d620538336&vid=4ac945220fa3730d357572c05c3735d620538336&prid=web&clid=SAPDAM