Octan-1-sulfonsyre natrium salt monohydrat ≥99.0% (by titrimetric analysis) for ion pair chromatography, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Sodium 1-octansulphonate monohydrate, Sodium 1-octanesulphonate monohydrate
74882-10G-F 74882-50G-F 74882-250G-F
74882-10G-FEA 1140 DKK
74882-10G-F 74882-50G-F 74882-250G-F
Octan-1-sulfonsyre natrium salt monohydrat ≥99.0% (by titrimetric analysis) for ion pair chromatography, Supelco®
Octan-1-sulfonsyre natrium salt monohydrat
Formel: C₈H₁₇NaO₃S·1H₂O
Molekylvægt: 234.29 g/mol
MDL Number: MFCD00007544
CAS-nummer: 207596-29-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dea818c4c50e33f0b0142cf2017a315735797dd5&vid=dea818c4c50e33f0b0142cf2017a315735797dd5&prid=web&clid=SAPDAM