2,2,3,3,3-Pentafluor-1-propanol ≥98.5% (by GC), LiChropur™ for GC derivatisation, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: 1H,1H-Pentafluoro-1-propanol, PFPOH (pentafluoropropanol), 1H,1H-Pentafluoropropanol-1, 2,2,3,3,3-Pentafluoropropanol
79879-10ML 79879-10X1ML
79879-10MLEA 1120 DKK
79879-10ML 79879-10X1ML
2,2,3,3,3-Pentafluor-1-propanol ≥98.5% (by GC), LiChropur™ for GC derivatisation, Supelco®
2,2,3,3,3-Pentafluoro-1-propanol
Formel: C₃H₃F₅O
MDL Number: MFCD00004673
CAS-nummer: 422-05-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e84b0588b7f0d36881c58108aba50aac351968e1&vid=e84b0588b7f0d36881c58108aba50aac351968e1&prid=web&clid=SAPDAM