Mini diaphragm vacuum pumps and compressors, airJet
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5625-2000 5625-2001
BURK5625-2000EA 11830 DKK
BURK5625-2000 BURK5625-2001
Mini diaphragm vacuum pumps and compressors, airJet
Pumper Membranpumper, vakuum
It is a single-head membrane vacuum pump which is widely used in research laboratories and industrial applications.
  • Membrane is made of EPDM (ethylene propylene diene monomer (M-class) rubber)
  • High level of gas tightness
  • Environment friendly
  • Maintenance-free
  • Low vibrations
  • 100% oil-free evacuation
Used for pumping, evacuate air, neutral or slightly aggressive or corrosive gases and vapours.
Leveringsinformation: Includes diaphragm pump with hose connector for hoses with 4 mm internal diameter.