Cytology funnels, EZ single Cytofunnel™, with white filter cards and caps
Leverandør: Epredia
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A78710003 A78710020
SHANA78710003EA 1050 DKK
SHANA78710003 SHANA78710020
Cytology funnels, EZ single Cytofunnel™, with white filter cards and caps
Tragte Cytologitragte
A disposable sample chamber with cap and pre-attached white filter card for high absorbency, this product is meant for single use with all cytocentrifuge applications.
  • Reduces the risk of exposure to pathogenic samples
  • Disposable to eliminate time-consuming decontamination and cleaning
  • Decreases the possibility of cross-contamination
  • Produces more cost-effective, thin-layer preparations when compared to other thin-layer methods
  • Holds sample volumes up to 0,5 ml
Optimise cell yield with these funnels.
Leveringsinformation: Includes closure caps, easy-to-use operational illustrations.