Poly(acrylic acid sodium salt) analytical standards, for GPC
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
81117-250MG 81123-250MG 81131-250MG
81117-250MGEA 10460 DKK
81117-250MG 81123-250MG 81131-250MG
Poly(acrylic acid sodium salt) analytical standards, for GPC
Standarder Kromatografi Standarder
Poly(acrylic acid sodium salt) is a hydrophilic polymer, which can find wide applications in the production of consumer product like diapers, because of its ability to absorb many more times its mass in water.
Certificeringer: Exact values can be taken from the accompanying certificate of analysis
Emballering: Bottomless glass bottle. Contents are inside inserted fused cone.
Advarsel: Limited shelf life, expiry date on the label
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9fb860850991f6dc6ef680869b487ef18f72efc2&vid=9fb860850991f6dc6ef680869b487ef18f72efc2&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d56cb53db550f37fcf8175f895e5ac0b38bb001&vid=7d56cb53db550f37fcf8175f895e5ac0b38bb001&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=770604bb5d2b68f7b15f1e32b4def5fd23b0595f&vid=770604bb5d2b68f7b15f1e32b4def5fd23b0595f&prid=web&clid=SAPDAM