Agarose LE, ultra-pure molecular bology grade
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
41028-25G 41028-100G 41028-500G
41028-25GEA 769 DKK
41028-25G 41028-100G 41028-500G
Agarose LE, ultra-pure molecular bology grade
Elektroforese geler Agarose og Akrylamid
High performance agarose for nucleic acid electrophoresis (analytical or preparative) and blotting. Validated for use with GelRed®, GelGreen®, and DNAzure® gel stains.
Ultra-pure LE agarose can be used for all routine electrophoresis applications. It has low electroendosmosis (EEO) which confers high electrophoretic mobility. These have high strength and shows low background after staining with common nucleic acid dyes.