Single channel micro pipette starter-sets
Leverandør: VITLAB
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
33331 33333 33332
613-2786EA 7930 DKK
613-2786 613-2788 613-2787
Single channel micro pipette starter-sets
Pipetter Automatiske pipetter
These starter sets includes three variable VITLAB® micropipettes with different volumes and associated, colour-coded tip boxes and three rack mounts for appropriate storage of micropipettes.
Certificeringer: Micropipettes are completely autoclavable at 121 °C (2 bar) according to DIN EN 285.
Leveringsinformation: Starter set 'Mini' includes VITLAB® micropipettes (0,5 to 10 μl, 10 to 100 μl, 100 to 1000 μl), tip-boxes (0,5 to 20 μl, 2 to 200 μl
, 50 to 1000 μl)
Starter set 'Classic' includes VITLAB® micropipettes (2 to 20 μl, 20 to 200 μl
, 100 to 1000 μl), tip-boxes (2 to 200 μl
(2x), 50 to 1000 μl)
Starter set 'Maxi' includes VITLAB® micropipettes (100 to 1000 μl, 500 to 5000 μl, 1000 to 10000 μl), tip-boxes (50 to 1000 μl, 0,5 to 5 ml, 1 to 10 ml)