Disinfectant spray, Contec® CyChlor
Leverandør: CONTEC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FBC500CC FBT100CC FBC100CC
CONTFBC500CCEA 1150 DKK
CONTFBC500CC CONTFBT100CC CONTFBC100CC
Disinfectant spray, Contec® CyChlor
Renseopløsninger Disinfektanter
Stabilised Hypochlorous acid with purified water. Contec CyChlor is a new broad spectrum disinfectant designed for every day use.
  • Broad spectrum efficacy in 3 mins saves time spent on biodecontamination
  • Filtered to 0,2 μm and filled in a Grade A environment ensuring the product is free from contamination and particulates
  • Contains no quaternary ammonium or surfactant so it's very low residue, saving time on residue removal
  • Trigger spray can be set to jet or spray - large droplet size reduces the risk of inhalation and provides good surface coverage
  • Not classed as corrosive and can be used safely in all areas of the cleanroom
No hazard classification so only basic PPE required and no special disposal is necessary.
Bestillingsoplysninger: A sterile version is available.
Leveringsinformation: Supplied as a 1L trigger spray or 1L capped container, or 5L capped containers for when larger volumes are required.
Emballering: Double bagged packed in linear tear packaging to aid the transfer of the product into a pharmaceutical cleanroom and also easy to open even when wearing gloves.
Advarsel: Use biocides safely. Always read the label and product information before use.