Portable balances, Compass™ CR series
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30428213 30428214 30428215 30428216
611-5259EA 352 DKK
611-5259 611-4792 611-5260 611-5261
Portable balances, Compass™ CR series
Vægte
The Compass™ CR series provides basic weighing functionality and competitive performance at an economical price. These portable scales have an ABS housing and pan, feature a slim, stackable design, and an enlarged weighing platform means they are ideal for a variety of portable weighing applications.
  • Practical design for convenient storage allows up to four scales to be stacked, saving valuable storage space when not in use
  • The enlarged weighing platform is ideal for use with a wide range of sample sizes
Bestillingsoplysninger: A range of optional accessories are available.