Methylenblåt for microscopy (C.I. 52015)
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue
3470.0025EA 463 DKK
3470.0025 3470.0100
Methylenblåt for microscopy (C.I. 52015)
Methylenblåt
Formel: C₁₆H₁₈ClN₃S
Molekylvægt: 319.86 g/mol
Smeltepunkt: 180
MDL Number: MFCD00150008
CAS-nummer: 61-73-4
Merck Index: 12,06137
Bestil nu
Specifikationer Testresultater
Proportion A635/A662 0.54 --> 0.62