Precision balances, portable, Compass™ CX series
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30428208 30428209 30428210 30428211 30428212
611-5254EA 611 DKK
611-5254 611-5255 611-5256 611-5257 611-5258
Precision balances, portable, Compass™ CX series
Vægte
Designed with energy efficiency in mind, the Compass CX series offers a battery life of up to 1000 continuous hours, allowing for uninterrupted operation. Compass™ CX scales have a stainless steel pan and are ideal for use in laboratory, industrial, education, and even household settings.
  • Streamlined two-button operation means that the balance can be operated easily
  • Large backlit LCD enables easy viewing of weighing results even in dim lighting conditions, and from a distance
  • Slim, light weight design for convenient storage allows up to four scales to be stacked, saving valuable storage space when not in use
  • Low power consumption design and alkaline batteries capable of up to 1000 hours of continuous use
Bestillingsoplysninger: A range of optional accessories are available.