Colorimetric rapid tests, copper, VISOCOLOR®
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
920050 920150 931037 931237
MANA920050EA 1710 DKK
MANA920050 MANA920150 MANA931037 MANA931237
Colorimetric rapid tests, copper, VISOCOLOR®
Miljø test kit
The VISOCOLOR® testkits are devided into VISOCOLOR® alpha, ECO and HE. Every product range offering its specific strenght to provide a tailored test for each and every user and application.
  • Portable
  • Environmentally friendly
  • Convenient handling
  • Application-oriented measuring ranges
In water copper(II) can be found in dissolved as well as in undissolved form. Copper(I) compounds and undissolved copper(II) compounds are not determined unless they are decomposed with concentrated nitric acid prior to the test.