Colorimetric rapid tests, iron, VISOCOLOR®
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
920040 920140 931026 931226 931025 931225
MANA920040EA 1710 DKK
MANA920040 MANA920140 MANA931026 MANA931226 MANA931025 MANA931225
Colorimetric rapid tests, iron, VISOCOLOR®
Miljø test kit
The VISOCOLOR® testkits are devided into VISOCOLOR® alpha, ECO and HE. Every product range offering its specific strenght to provide a tailored test for each and every user and application.
  • Portable
  • Environmentally friendly
  • Convenient handling
  • Application-oriented measuring ranges
Natural waters as well as waste waters often contain some iron. It can be present as Fe(II) or Fe(III) ions if the pH value of the water is below 3 or if the water is free of oxygen. At higher pH values Fe(III) forms an insoluble oxyhydrate. Frequently the sample is in a transition state with finely dispersed iron oxides. In waste water and natural water containing humic acids iron is often present in the form of a complex salt. The VISOCOLOR® test kits only determine iron which is present in dissolved form as Fe²⁺ or Fe³⁺ ions. Iron complexes are not covered unless they are decomposed by oxidation with nitric acid and sulphuric acid.
10
Iron