Colorimetric rapid tests, ammonium, VISOCOLOR®
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
920006 920106 931008 931208 935012 931010 931210
MANA920006EA 1710 DKK
MANA920006 MANA920106 MANA931008 MANA931208 MANA935012 MANA931010 MANA931210
Colorimetric rapid tests, ammonium, VISOCOLOR®
Miljø test kit
The VISOCOLOR® testkits are devided into VISOCOLOR® alpha, ECO and HE. Every product range offering its specific strenght to provide a tailored test for each and every user and application.
  • Portable
  • Environmentally friendly
  • Convenient handling
  • Application-oriented measuring ranges
Ammonium ions occur primarily in domestic waste waters, but frequently also in industrial waste waters. In surface and ground waters ammonium ions indicate decomposition of animal or vegetable substances. Control of the ammonium values is therefore important to the water supply.