SPE columns, Supra-Clean® Florisil (FL-S) Pesticide grade
Leverandør: PerkinElmer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
N9306443 N9306447 N9306436 N9306470 N9306516
PERCN9306443EA 1400 DKK
PERCN9306443 PERCN9306447 PERCN9306436 PERCN9306470 PERCN9306516
SPE columns, Supra-Clean® Florisil (FL-S) Pesticide grade
Kromatografikolonner
Supra-Clean® Florisil Pesticide (FL-S) SPE columns are recommended for extracting polar compounds when analysing pesticides.
  • Hydrophilic mechanism
  • Ideal for polar compounds in non-polar matrix
  • pH range: 2 to 8
Alternative to silica for viscous matrices.