Electrical insulation tape, vinyl, Scotch® Super 33+™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
331933 332533 335033A
MMMA331933EA 9300 DKK
MMMA331933 MMMA332533 MMMA335033A
Electrical insulation tape, vinyl, Scotch® Super 33+™
Tape
Electric tape Scotch Super 33+ is a particularly elastic, UV-resistant premium tape with very good electrical properties and resistance to moisture, alkalis, many acids and solvents. Best winding properties even at low temperatures.
  • Pressure-sensitive rubber based adhesive
  • Excellent resistance to oil/solvents
  • Self-extinguishing and no self-welding
  • Electrical insulation for applications up to 1 kV
  • Inhibits corrosion of electrical conductors
  • Compatible with solid dielectric cable insulations
  • Compatible with rubber and synthetic splicing compounds, as well as epoxy and polyurethane resins
Vinyl Electrical Tape is a premium grade, 7 mil thick, all-weather vinyl-insulating tape. It is designed to perform continuously in ambient temperatures up to 105 °C (220 °F). The tape is conformable for cold weather application down to –18 °C (0 °F). The combination of elastic backing and aggressive adhesive provides moisture-tight electrical and mechanical protection with minimum bulk.
It is used as primary electrical insulation for all wire and cable splices rated up to 600 V and up to 105 °C (220 °F) and for 600 V bus applications, and protective jacketing for low and high voltage bus. Protective jacketing for high voltage cable splices and repair, harnessing of wires and cables.