VWR®, Microcentrifuge Tubes with Flat Screw Caps
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
525-1137EA 354 DKK
525-1137 525-1138 525-1139 525-1140 525-1141 525-1143 525-1144 525-1145 525-1146 525-1147 525-1148 525-1149 525-1155 525-1156 525-1157 525-1158 525-1159 525-1160
VWR®, Microcentrifuge Tubes with Flat Screw Caps
Rør Mikrocentrifugerør
These tubes are made from ultra-clear USP Class VI PP that allows easy viewing of contents.
  • RNase-, DNase- and endotoxin-free
  • Engraved graduations ensure accuracy
  • Fit most common rotors
  • Smooth surface for applying labels
  • Sterile tubes are sterilised by irradiation
These tubes feature universal screw threads for use with most popular brand screw caps. Low profile caps include a silicone rubber O-ring, ensuring a leakproof seal. (Please NOTE: Silicone O-rings have been used from June 22, 2022, first lot number: 220622626D. Manufacturer stopped using the MCT cap with ethylene propylene O-ring after June 22, 2022.)
Tubes and caps are autoclavable at 121 °C and freezable to -80 °C.
Certificeringer: Manufactured in a Class 100 000 cleanroom environment.
Manufactured under ISO 13485:2016 and ISO 9001:2015 quality management systems.
Certificate available for every lot.
Leveringsinformation: Tubes are packed 500 per bag. Assorted pack of caps (525-1187) contains equal quantities of red, orange, yellow, blue and green.