ESD safety shoes, lace-up, uvex 2 xenova®, 9555
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9555.2/48 9555.2/49 9555.2/50 9555.2/51 9555.2/52 9555.3/38 9555.3/39 9555.3/40 9555.3/41 9555.3/42 9555.3/43 9555.3/44 9555.3/45 9555.3/46 9555.3/47 9555.3/48 9555.3/49 9555.3/50 9555.3/51 9555.3/52 9555.1/38 9555.1/39 9555.1/40 9555.1/41 9555.1/42 9555.1/43 9555.1/44 9555.1/45 9555.1/46 9555.1/47 9555.1/48 9555.1/49 9555.1/50 9555.1/51 9555.1/52 9555.2/38 9555.2/39 9555.2/40 9555.2/41 9555.2/42 9555.2/43 9555.2/44 9555.2/45 9555.2/46 9555.2/47
UVEX9555.2/48EA 849 DKK
UVEX9555.2/48 UVEX9555.2/49 UVEX9555.2/50 UVEX9555.2/51 UVEX9555.2/52 UVEX9555.3/38 UVEX9555.3/39 UVEX9555.3/40 UVEX9555.3/41 UVEX9555.3/42 UVEX9555.3/43 UVEX9555.3/44 UVEX9555.3/45 UVEX9555.3/46 UVEX9555.3/47 UVEX9555.3/48 UVEX9555.3/49 UVEX9555.3/50 UVEX9555.3/51 UVEX9555.3/52 UVEX9555.1/38 UVEX9555.1/39 UVEX9555.1/40 UVEX9555.1/41 UVEX9555.1/42 UVEX9555.1/43 UVEX9555.1/44 UVEX9555.1/45 UVEX9555.1/46 UVEX9555.1/47 UVEX9555.1/48 UVEX9555.1/49 UVEX9555.1/50 UVEX9555.1/51 UVEX9555.1/52 UVEX9555.2/38 UVEX9555.2/39 UVEX9555.2/40 UVEX9555.2/41 UVEX9555.2/42 UVEX9555.2/43 UVEX9555.2/44 UVEX9555.2/45 UVEX9555.2/46 UVEX9555.2/47
ESD safety shoes, lace-up, uvex 2 xenova®, 9555
Sko og støvler
Very lightweight water repellent uvex 2 xenova® shoes are made from full-grain smooth leather upper, soft padded collar and a dual-density slip-resistant Xenova® sole resistant to heat, chemicals, oils and gasoline. All sole materials are free of silicones, plasticisers and other paint-wetting impairment substances and suitable for people allergic to chrome.
  • Compact design, anatomical shape, good lateral stability, no thermal conductivity, non-magnetic
  • Anti-twist rear cap provides more stability and kink protection
  • Climazone technology for improved comfort
  • Anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Heat-resistant loop lacing system
  • DUO-PUR sole, non-marking, with excellent cushioning and abrasion-resistant outsole
  • Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
  • Shock absorbing heel and forefoot
  • Metal-free and penetration-resistant uvex xenova® midsole
  • uvex xenova® composite toe cap (200 J)
  • Fulfils the ESD requirement: Discharge resistance <35 megohm
Used for medium-duty applications.
Colour: Blue/black
Certificeringer: EN 61340 (ESD), EN ISO 20345:2011; S3, SRC