Tin(II)chlorid dihydrat 98% ACS, Fluka™
Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Stannous chloride dihydrate
243523-2.5KG 243523-12KG
243523-2.5KGEA 6800 DKK
243523-2.5KG 243523-12KG
Tin(II)chlorid dihydrat 98% ACS, Fluka™
Tin(II)chlorid dihydrat
Formel: SnCl₂·2H₂O
Molekylvægt: 225.65 g/mol
Kogepunkt: 623 Dec. (on heating)
Smeltepunkt: 37…38 °C
Massefylde: 2.71 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149863
CAS-nummer: 10025-69-1
EINECS: 231-868-0
UN nummer: 3260
ADR: 8 III
Merck Index: 13,08861
Bestil nu
Specifikationer Testresultater
Assay 98.0 - 103.0 %
Solubility in HCl complying
Calcium (Ca) Max. 0.005 %
Iron (Fe) Max. 0.003 %
Potassium (K) Max. 0.005 %
Sodium (Na) Max. 0.01 %
Lead (Pb) Max. 0.01 %
Sulfate (SO₄) Max. 0.003 %