ESD safety shoes, lace up, Wellmaxx, Travis low
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
729820/37 729820/38 729820/39 729820/40 729820/41 729820/42 729820/43 729820/44 729820/45 729820/46 729820/47 729820/48 729820/36
ELTN729820/37EA 694 DKK
ELTN729820/37 ELTN729820/38 ELTN729820/39 ELTN729820/40 ELTN729820/41 ELTN729820/42 ELTN729820/43 ELTN729820/44 ELTN729820/45 ELTN729820/46 ELTN729820/47 ELTN729820/48 ELTN729820/36
ESD safety shoes, lace up, Wellmaxx, Travis low
Sko og støvler
ESD safety shoes made from hydrophobised microfibre and hydrophobised textile material, a full padded bellows tongue and a PU/PU sole WELLMAXX TRAINERS.
  • Fabric lining for natural ventilation
  • Full-length inlay sole ESD PRO black
  • Soft fleece ESD insole
  • Protective composite toecap (200 J)
  • Sole core made of Infinergy® by BASF
  • Metal-free and leather-free equipment
  • Anatomical footbed gives the support and stability
  • Reflective material increases safety
Ideal for use in areas where exposure to moisture is expected.
Colour: Grey/red
Certificeringer: EN 61340-5-1 (ESD); EN ISO 20345; S2, SRC, form A