VWR®, renrumsstrømper, ESD, engangs
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
113-9409EA 10820 DKK
113-9409 113-9410
VWR®, renrumsstrømper, ESD, engangs
Engangsbeklædning
These disposable socks are made of polyester filament yarn and anti-static carbon fibre.
  • Ultra-clean treatment
  • Low particle generation
  • Friction resistance
  • Soft and comfortable to wear
  • ESD properties
Suitable for use in ISO Class 4 to 6 (Class 10 to 1000) cleanrooms.