ESD safety shoes, lace-up, Landon Low
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
729505/40 729505/41 729505/42 729505/43 729505/44 729505/45 729505/46 729505/47 729505/48
ELTN729505/40EA 634 DKK
ELTN729505/40 ELTN729505/41 ELTN729505/42 ELTN729505/43 ELTN729505/44 ELTN729505/45 ELTN729505/46 ELTN729505/47 ELTN729505/48
ESD safety shoes, lace-up, Landon Low
Sko og støvler
Metal- and leather-free ESD safety shoes made from textile material/microfibre, padded tongue and a dual-density TPU/PU sole MAXXIMO.
  • Fabric lining for natural ventilation
  • Full-length inlay sole SPORTIVE ESD black
  • Soft fleece ESD insole
  • Protective steel toecap (200 J)
  • Sole core made of Infinergy® by BASF
  • Anatomically shaped insole and ankle support for improved stability
Ideal for use in dry work areas such as industry, storage, transport, or assembly.
Colour: Grey
Certificeringer: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1, SRC, form A