Test strips, total iron, QUANTOFIX®
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
91330 91344
MANA91330EA 410 DKK
MANA91330 MANA91344
Test strips, total iron, QUANTOFIX®
Miljø test kit
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. The test strips are intended for single use, maintenance or calibration are not required.
  • Results within seconds: Just dip, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips allow the quick and easy determination of total iron in solutions. The easy dip-and-read-procedure provides reliable results without the need for training or experience. Within one minute one gets reliable results.
Iron is frequently used for water pipes and vessels in industrial applications. Therefore, dissolved iron in process water is an important indicator for the level of corrosion. In drinking water, dissolved iron is not desirable as it leads to a brown color and a foul taste. These test strips are the ideal tool to test iron concentrations in water quickly and easily down to low concentrations.