ESD safety shoes, lace-up, Officer XXB Low
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
72302/47 72302/40 72302/41 72302/42 72302/43 72302/44 72302/45 72302/46 72302/48
ELTN72302/47EA 768 DKK
ELTN72302/47 ELTN72302/40 ELTN72302/41 ELTN72302/42 ELTN72302/43 ELTN72302/44 ELTN72302/45 ELTN72302/46 ELTN72302/48
ESD safety shoes, lace-up, Officer XXB Low
Sko og støvler
Metal-free business ESD safety shoes made from cowhide leather, padded tongue plus a mono-density PU sole (Wellmaxx Business).
  • Leather lining
  • Full length inlay sole (Sportive ESD), grey
  • Protective steel toecap (200 J)
  • Sole core made of Infinergy® by BASF
  • Soft fleece ESD insole
Ideal for use in areas where exposure to moisture is expected.
Colour: Black
Certificeringer: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S2, SRC, form A