Conductivity standards, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
COND147-1L COND50-500ML COND1412-1L COND1412-500ML COND147-500ML COND25-500ML COND75-500ML COND2060-500ML
COND147-1LEA 783 DKK
COND147-1L COND50-500ML COND1412-1L COND1412-500ML COND147-500ML COND25-500ML COND75-500ML COND2060-500ML
Conductivity standards, Supelco®
Standarder Ledningsevne standarder
100 µmhos/cm = 100 µS/cm
Certificeringer: These certified reference materials (CRMs) ares produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.