Analytical balances, PR series
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30430068 30430067 30524384 30430070 30430069 30524385
611-5035EA 9820 DKK
611-5035 611-5036 611-4965 611-5024 611-5026 611-4964
Analytical balances, PR series
Vægte
The PR series of analytical balances has been designed for routine weighing applications in the workplace. The durable design has a stainless steel weighing pan for long-term use. Featuring RS232 connectivity for easy communication, an easy-to-read, bright backlit display and a simple interface for uncomplicated operation.
  • Three essential weighing modes
  • Choice of models with internal or external calibration
  • Balances are easy to operate, with almost no training required
  • Small footprint saves bench space, while providing a large weighing surface
  • Applications
More Product Information