S-Nitrosoglutathione (GSNO) ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Leverandør: Merck Millipore Calbiochem
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-(nitrosothio)-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, GSNO, Nitrosoglutathione, S-Nitroso-L-Glutathione
Packaged under inert gas.
487920-10 487920-50 487920-1
487920-10EA 1330 DKK
487920-10 487920-50 487920-1
S-Nitrosoglutathione (GSNO) ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
S-Nitrosoglutathione (GSNO)
A utility carrier of nitric oxide.
Breaks down at physiological pH to produce nitric oxide. Causes smooth muscle relaxation and inhibits platelet aggregation. Induces apoptosis in mouse thymocytes.
Formel: C₁₀H₁₆N₄O₇S
Molekylvægt: 336.319 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD00214341
CAS-nummer: 57564-91-7
Bestil nu
Specifikationer Testresultater
Purity (HPLC) ≥95%