Furaptra (Mag-Fura-2) AM Ester ≥95% (by TLC) fluorescent indicator, for calcium determination
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Mag-Fura-2 AM ester
50037 50039 50038
50037.EA 3490 DKK
50037. 50039. 50038.
Furaptra (Mag-Fura-2) AM Ester ≥95% (by TLC) fluorescent indicator, for calcium determination
Furaptra (Mag-Fura-2) AM Ester
Furaptra, AM ester is a membrane-permeant form of Furaptra (Mag-Fura-2). The indicator is loaded into cells via simple incubation
  • Absorption/Emission 366nm/475nm (EtOAc)
  • Extinction Coefficient 31,000
  • Cell permeability: Membrane permeant
  • Colors: Green
  • Light yellow solid soluble in DMSO
  • Store at -20°C and protect from light
Formel: C₃₀H₃₀N₂O₁₉
Molekylvægt: 723 g/mol
Opbevaringstemperatur: Fryser
CAS-nummer: 130100-20-8