Wipes for general applications, J-Cloth
Leverandør: PGI NONWOVEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7450100 7450200 7450300 7450400 74505
115-2503EA 162 DKK
115-2503 115-2504 115-2505 115-2506 115-2507
Wipes for general applications, J-Cloth
Papirservietter Genanvendelige rengøringsklude Klude til alment brug
These wipes feature unique, open, wavy texture that enables efficient pick-up of any dirt and makes it easy to rinse, ensuring longer freshness.
  • Compostable
  • Granted with compostability logo
  • Open weave structure
  • Cleaning agent compatible
  • Five different colours
Certificeringer: FCC approved.