Prepaid subscription cards with activation code for SenseAnywhere data loggers
Leverandør: SenseAnywhere
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0630125976968 0630125976975 0630125976982 0630125976999
620-2410EA 689 DKK
620-2410 620-2428 620-2429 620-2430
Prepaid subscription cards with activation code for SenseAnywhere data loggers
Software Laboratoriesoftware
Top half prepaid subscription card
This card represents a number of Credits only valid for the SenseAnywhere portal
One Credit represents a one-year subscription for one sensor or 40 SMS notification messages
For more details see Help > Prepaid card once you have logged-in
Bottom half prepaid subscription card
1. Remove scratch-off paint from area above
2. Go to www.saclient.com/redeem
3. Log-in or create a new account
4. Follow instructions on the screen