Precision balances, Cubis® II high capacity
Leverandør: Sartorius Balances
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MCA12201S0-CEU MCA12201S0-S00 MCA20201S0-CEU MCA20201S0-S00 MCA32202P0-S00 MCA36201S0-CEU MCA36201S0-S00 MCA70201S0-CEU MCA70201S0-S00 MCE12201S0-CEU MCE12201S0-S00 MCE32202P0-S00 MCE36200S0-CEU MCE36200S0-S00 MCE36201S0-CEU MCE36201S0-S00
611-4685EA 69900 DKK
611-4685 611-4686 611-4697 611-4698 611-4720 611-4725 611-4726 611-4747 611-4748 611-4753 611-4754 611-4774 611-4777 611-4778 611-4779 611-4780
Precision balances, Cubis® II high capacity
Vægte
The Cubis® II range has a modular design that to allows the customer to choose the components that suit the application area that is of interest. For the customer, this configurable concept delivers quality weighing, together with a high degree of flexibility, without having to invest in expensive equipment that has features that may never be required.
  • Quick and easy motorised automatic levelling, for weighing modules up to 8,2 kg, units above 8,2 kg have manual levelling
  • Gesture control sensor recognises four gestures, it allows user to send a document to a device
  • Connectivity package links to diverse laboratory information systems (Windows File Server (SMB), FTP, FTPS and STARLIMS) without the need for additional software
  • Full data integrity following ALCOA+ principles, the balances audit trail is a tamper protected, time stamped on an electronic log file that is traceable to UTC and the balance contains an Alibi memory
Two different user interfaces are available to cover all facets of laboratory applications and enable a range of options for balance operation. The customer can then add the appropriate weighing platform to the choice of control unit.
MCE Essential User Interface: This is a large, high contrast touch display with factory installed essential weighing applications for easy operation.
MCA Advanced User Interface: This is a large 7" colour touch display featuring touch, scroll and swipe functionality with factory installed essential weighing applications and diverse QApp packages for optional software extension. An optional climate sensor which monitors the influencing environmental variables ensuring temperature, pressure and humidity do not negatively affect results is available. The Status Center shows all information about the balance and environmental conditions (calibration, levelling, temperature, humidity, air pressure and service) in a dashboard, giving detailed help and support in the event of warnings or errors.
Various Q-Apps, customer-specific applications, can be integrated into the balance. The applications are used to create customer-specific weighing tasks. These are reloadable application programs that guide the user step-by-step through the specific workflow, guaranteeing that the procedures described in the corresponding SOPs are observed at all times. As individual Q-Apps are performed according to their specific application, a variety of solutions for differential weighing, formula and filling quantity control applications are available as Q-App packages. Workflows can be performed independently and do not necessarily require a PC connection.
Certificeringer: Supports all requirements for FDA directive 21 CFR part II and EU Annex II compliance.
Leveringsinformation: There are different optional software packages, each contains applications or functions to enhance and customise the balance for the users’ workflows. Choice of QApp Package: Pharma; Advanced Applications: Utilities; Connectivity or the all inclusive QApps package includes all of these 4 packages. (denoted by QP99)
Advarsel: Note: Models with suffix -CEU are verified version Europe (except FR) and -S00 suffix denotes standard version worldwide.