Analytical balances, Cubis® II
Leverandør: Sartorius Balances
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MCA124SI-CEU MCA124SI-S00 MCA124SU-CEU MCA124SU-S00 MCA224SI-CEU MCA224SI-S00 MCA224SU-CEU MCA224SU-S00 MCA324SI-CEU MCA324SI-S00 MCA324SU-CEU MCA324SU-S00 MCA524SI-CEU MCA524SI-S00 MCA524SU-CEU MCA524SU-S00 MCE124SU-S00 MCE224SU-CEU MCE224SU-S00 MCE324SU-CEU MCE324SU-S00
611-4687EA 118750 DKK
611-4687 611-4688 611-4689 611-4690 611-4705 611-4706 611-4707 611-4708 611-4721 611-4722 611-4723 611-4724 611-4733 611-4734 611-4735 611-4736 611-4756 611-4765 611-4766 611-4775 611-4776
Analytical balances, Cubis® II
Vægte
The Cubis® II range has a modular design that to allows the customer to choose the components that suit the application area that is of interest. For the customer, this configurable concept delivers quality weighing, together with a high degree of flexibility, without having to invest in expensive equipment that has features that may never be required.
  • Choice of draft shield: Manual glass (U), motorised (A) or motorised with built-in de-ioniser for effective removal of static charges (I)
  • Quick and easy motorised automatic levelling
  • Gesture control sensor recognises four gestures, it allows user to open or close the automatic draft shield, start the ioniser or send a document to a device
  • Connectivity package links to diverse laboratory information systems (Windows File Server (SMB), FTP, FTPS and STARLIMS) without the need for additional software
  • Built in ioniser removes electrostatic charges at the press of a key or automatically when draft shield closes
  • Full data integrity following ALCOA+ principles, the balances audit trail is a tamper protected, time stamped electronic logfile traceable to UTC and the balance contains an Alibi memory
Two different user interfaces are available to cover all facets of laboratory applications and enable a range of options for balance operation. The customer can then add the appropriate weighing platform to the choice of control unit. For these analytical balances the choices are, not only the control unit and balance capability, but the type of draft shielding that will benefit the application. Choice of automatic glass draft shield with or without integrated ionizer.
MCE Essential User Interface: This is a large, high contrast touch display with factory installed essential weighing applications for easy operation.
MCA Advanced User Interface: This is a large 7" colour touch display featuring touch, scroll and swipe functionality with factory installed essential weighing applications and diverse QApp packages for optional software extension. An optional climate sensor which monitors the influencing environmental variables ensuring temperature, pressure and humidity do not negatively affect results is available. The status center shows all information about the balance and environmental conditions (calibration, levelling, temperature, humidity, air pressure and service) in a dashboard, giving detailed help and support in the event of warnings or errors.
Various Q-Apps, customer-specific applications, can be integrated into the balance. The applications are used to create customer-specific weighing tasks. These are reloadable application programs that guide the user step-by-step through the specific workflow, guaranteeing that the procedures described in the corresponding SOPs are observed at all times. As individual Q-Apps are performed according to their specific application, a variety of solutions for differential weighing, formula and filling quantity control applications are available as Q-App packages. Workflows can be performed independently and do not necessarily require a PC connection.
Certificeringer: Supports all requirements for FDA directive 21 CFR part II and EU Annex II compliance.
Bestillingsoplysninger: There are different optional software packages, each contains applications or functions to enhance and customise the balance for the users’ workflows. Choice of QApp package: Pharma; Advanced applications: Utilities; Connectivity or the all inclusive QApps package includes all of these 4 packages.
Advarsel: Note: Models with suffix -CEU are verified version Europe (except FR) and -S00 suffix denotes standard version worldwide.