Alcohol metabolite Standards, ethyl sulphate sodium salt, CRM, Cerilliant®, Supelco®
Leverandør: MerckSamlet vurdering
Gennemsntilig vurdering
Ethyl sulfate is a primary urinary metabolite of consumed alcohol and is indicative of recent alcohol consumption.
- Snap-N-Spike®/Snap-N-Shoot®
Certified Spiking Solution® suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine alcohol testing.
Advarsel: Storage temperature: −20 °C (freezer).