Storage bottles, octogonal, PET, Corning® Gosselin™
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
P1000B-50 P125B-50 P250B-50 P500B-50 P1000B-06 P125B-08
GOSSP1000B-50EA 2480 DKK
GOSSP1000B-50 GOSSP125B-50 GOSSP250B-50 GOSSP500B-50 GOSSP1000B-06 GOSSP125B-08
Storage bottles, octogonal, PET, Corning® Gosselin™
Flasker Plastflasker
These storage bottles are made from break resistant polyethylene terephthalate (PET) with transparency equivalent to glass.
  • Molded graduations
  • Ergonomic shape for easy handling
  • Leak proof, tamper evident HDPE screw cap
Available with Sterility Assurance Level (SAL) of 10⁻³.
With wide mouth opening and designed to collect samples or store laboratory fluids.