VWR®, IOS LWD U-Plan Objectives for Fluorescence Applications, F Series
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
630-2708EA 11940 DKK
630-2708 630-2709 630-2706 630-2707
VWR®, IOS LWD U-Plan Objectives for Fluorescence Applications, F Series
Mikroskop mål
These objectives are designed for use with inverted microscopes. They were specifically developed for high class performance and contrast required by the most demanding users.
  • Very high resolution and contrast: Ideal for UV fluorescence applications
  • Designed to ensure field flatness up to F.N. 25.
  • Long Working Distance provides a wider working space between the lens surface and the object
  • Plan-Fluorite (or Semi-Apochromatic) design ensures additional spherical aberration correction for superior resolution and greater numerical apertures