Leather gloves, uvex top grade 8000
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6029509 6029510 6029511
UVEX6029509EA 605 DKK
UVEX6029509 UVEX6029510 UVEX6029511
Leather gloves, uvex top grade 8000
Handsker
These safety gloves are made of extremely durable leather with breathable cotton on the back of the hand.
  • Fingertip, wrist and knuckle protection
  • Good mechanical abrasion resistance
  • Exceptional grip on dry and (slightly) damp tools
  • Excellent wearer comfort
The glove is constructed of double-stitched seams, rubberised cuff and full-grain leather palm, index finger, fingertips, knuckle trim and thumb.
Typical applications include: Assembly, inspection, light to medium metal processing and manual work.
Certificeringer: EN 388:2016 (4144X)